Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF-1 Mouse anti-Human, Mouse, Clone: REMB6, Exalpha Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$332.32 - $522.16

Specifications

Antigen LEF-1
Clone REMB6
Concentration 0.5-2 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-1847 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
T105M
250 μg
Each for $522.16
Add to cart
 
50-210-1848 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
T105MS
100 μg
Each for $332.32
Add to cart
 
Description

Description

LEF-1 Monoclonal antibody specifically detects LEF-1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications

Specifications

LEF-1
0.5-2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.08% sodium azide; pH
3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; LEF-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; Lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3
Full-length recombinant LEF-1 protein
Monoclonal
Protein A/G
Antibody
REMB6
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human, Mouse
P27782, Q9UJU2
16842, 51176
IgG1
Primary
-20° C, Avoid Freeze/Thaw Cycles
LEF1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit