Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Mouse anti-Human, Clone: 1C3.1D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266624

Catalog No. NBP266624


Add to cart

Description

Description

LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

LEF1
1C3.1D10
Western Blot
Purified
LEF1
Recombinant full length LEF-1 protein
Protein A or G purified
DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary
51176
Western Blot
Unconjugated
PBS with 0.08% Sodium Azide
DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Mouse
IgG1
100μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit