Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEF1 Mouse anti-Human, Clone: REMB6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266626
Description
LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
LEF1 | |
REMB6 | |
Western Blot, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence | |
Purified | |
LEF1 | |
Full-length recombinant LEF-1 protein | |
Protein A or G purified | |
DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway | |
Primary | |
51176 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA | |
Mouse | |
IgG1 | |
250μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Monkey, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title