Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Rabbit anti-Human, Clone: LEF1/422R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen LEF1
Clone LEF1/422R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123040
missing translation for 'viewDocuments'
Novus Biologicals
NBP30778320UG
20 μg
Each for $159.82
Add to cart
 
NB123039
missing translation for 'viewDocuments'
Novus Biologicals
NBP30778310UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

LEF1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Synthetic peptide corresponding to residues within aa100-200 of LEF1
Monoclonal
Protein A or G purified
LEF1/422R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
51176
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit