Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leflunomide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
75706-12-6
C12H9F3N2O2
270.211
VHOGYURTWQBHIL-UHFFFAOYSA-N
leflunomide,arava,leflunomidum,lefunamide,leflunomida,leflunomidum inn-latin,5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide,leflunomida inn-spanish,su 101 pharmaceutical,arava tn
3899
CHEBI:6402
5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F
This item is not returnable. View return policy
75706-12-6
C12H9F3N2O2
270.211
VHOGYURTWQBHIL-UHFFFAOYSA-N
leflunomide,arava,leflunomidum,lefunamide,leflunomida,leflunomidum inn-latin,5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide,leflunomida inn-spanish,su 101 pharmaceutical,arava tn
3899
CHEBI:6402
5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F

Inhibitor of DHODH

Inhibitor of dihydroorotate dehydrogenase (IC50 = 2.5 μM). Inhibits de novo pyrimidine synthesis in human T cells in vitro; also inhibits lymphocyte proliferation. Exhibits efficacy in several animal models of autoimmune disease, arthritis and graft rejection. Active metabolite, teriflunomide (A77 1726, Cat. No. 5069), also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 270.21
Percent Purity >99%
Chemical Name or Material Leflunomide

Recommended Storage : Store at +4°C