Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lefty-1 Rat anti-Mouse, Clone: RM0121-8B65, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP122499
Description
Lefty-1 Monoclonal antibody specifically detects Lefty-1 in Mouse samples. It is validated for Western Blot.Specifications
Lefty-1 | |
RM0121-8B65 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
AI450052, EBAF, Leftb, lefty, lefty-1, Stra3, TGFB4 | |
Rat | |
IgG2 | |
0.1 mg | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
Monoclonal | |
This was produced from a hybridoma (mouse myeloma0 fused with spleen cells from a rat immunized with mouse recombinant protein of Lefty-1. | |
Mouse |
Western Blot | |
LYOPH | |
Western Blot 1:500-1:1000 | |
Purified | |
LEFTY1 | |
Mouse recombinant protein of Lefty-1 | |
Protein G purified | |
RUO | |
Primary | |
10637 | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only