Learn More
Lefty-1 Rat anti-Mouse, DyLight 350, Clone: RM0121-8B65, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP122499UV
Catalog No. NBP122499UV
Add to cart
Description
Description
Lefty-1 Monoclonal antibody specifically detects Lefty-1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Lefty-1
RM0121-8B65
Western Blot
Purified
LEFTY1
Mouse recombinant protein of Lefty-1
Protein G purified
Store at 4C in the dark.
Monoclonal
This was produced from a hybridoma (mouse myeloma0 fused with spleen cells from a rat immunized with mouse recombinant protein of Lefty-1.
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
AI450052, EBAF, Leftb, lefty, lefty-1, Stra3, TGFB4
Rat
IgG2
0.1 ml
Primary
10637
Mouse