Learn More
National Safety Apparel™ Legging 5 oz. Carbon Kevlar Lined With 6 oz. Omni Quilted
SDP
Catalog Number p-7220290
Click to view available options
Size:
Large
X-Large
2X-Large
Size Large
Color Olive Green
Ankle Style Open
Seam Type Stitched
Certifications/Compliance Arc Rating: 40 cal/cm², CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Suspender Adjustable