Learn More
Legumain/Asparaginyl Endopeptidase Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 13 publications
Supplier:  R&D Systems AF2199SP
Catalog No. AF2199SP
Add to Cart
This item is not returnable. View return policy
Description
Description
Legumain/Asparaginyl Endopeptidase Polyclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
Legumain/Asparaginyl Endopeptidase
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Goat
Affinity Purified
RUO
5641
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q99538
LGMN
Mouse myeloma cell line NS0-derived recombinant human Legumain/Asparaginyl Endopeptidase Ile18-Tyr433 Accession # Q99538
25 μg
Primary
Detects human Legumain/Asparaginyl Endopeptidase in direct ELISAs and Western blots.
Human
IgG