Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legumain/Asparaginyl Endopeptidase Mouse anti-Human, Alexa Fluor™ 750, Clone: 46I3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221996AF750
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA.Specifications
Legumain/Asparaginyl Endopeptidase | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
LGMN | |
Human recombinant Legumain/Asparaginyl Endopeptidase | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, ELISA | |
46I3 | |
Western Blot, ELISA | |
AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
Mouse | |
Protein G purified | |
Cancer | |
5641.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title