Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Leishman Stain

Manufacturer:  Alfa Aesar™ A1164422

Quantity 100g

 View more versions of this product

Catalog No. AAA1164422


  Check Availability
Add to cart

Specifications

Description

Applications
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.

Solubility
Soluble in water (4 mg/ml), methanol (1 mg/ml), ethanol (4 mg/ml), and methyl cellosolve (20 mg/ml).

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Description & Specifications

Chemical Identifiers

12627-53-1
MFCD00131498
Leishman Stain

Specifications

12627-53-1
Odorless
Leishman Stain
MFCD00131498
100g
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit