Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEMD2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | LEMD2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170598
|
Novus Biologicals
NBP170598 |
100 μL |
Each of 1 for $436.00
|
|
Description
LEMD2 Polyclonal specifically detects LEMD2 in Human samples. It is validated for Western Blot.Specifications
LEMD2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
221496 | |
Synthetic peptides corresponding to LEMD2(LEM domain containing 2) The peptide sequence was selected from the middle region of LEMD2. Peptide sequence QDMERYPYVGILHVRDSLIPPQSRRRMKRVWDRAVEFLASNESRIQTESH. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
LEM domain containing 2 | |
LEMD2 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title