Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

LEMD3 Mouse anti-Human, Canine, Monkey, PE/Atto594, Clone: OTI6C10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271744PEATT594

Catalog No. N271744P594



Description

Description

LEMD3 Monoclonal antibody specifically detects LEMD3 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

LEMD3
OTI6C10
Flow Cytometry
Affinity Purified, Conjugated
LEMD3
Full length recombinant protein of human LEMD3 (NP_055134) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
inner nuclear membrane protein Man1, integral inner nuclear membrane protein, LEM domain containing 3, MAN1LEM domain-containing protein 3
Mouse
IgG1
0.1 mL
Primary
23592.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit