Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEMD3 Mouse anti-Human, Clone: OTI4E1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA500797
Description
Can function as a specific repressor of TGF-beta, activin, and BMP signaling through its interaction with the R-SMAD proteins. Antagonizes TGF-beta-induced cell proliferation arrest.Specifications
LEMD3 | |
Monoclonal | |
Unconjugated | |
LEMD3 | |
Liquid | |
LEMD3 | |
Full length human recombit protein of human LEMD3 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
OTI4E1 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y2U8 | |
MAN1 | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
23592 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title