Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lemon Flavor Extract, Pure, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle

L1180

Spectrum™ Lemon Flavor Extract, Pure
Packaging Amber Glass Bottle
Quantity 500 mL
CAS Max % 1
UN Number UN1197