Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lemon Oil, California Type, Cold-pressed, FCC, 2.2-3.8%, Spectrum™

Click to view available options
Quantity:
500 mL
15 kg
Packaging:
Amber Glass Bottle
Steel Pail
8008-56-8
8008-56-8

L1200, 8008-56-8

Spectrum™ Lemon Oil, California Type, Cold-pressed, FCC is used in aromatherapy and as a flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Steel Pail
Refractive Index 1.473-1.476
Quantity 15 kg
CAS Max % 1
UN Number UN1197
Percent Purity 2.2 to 3.8%