Learn More
Cayman Chemical LenalIdomIde 1g
SDP
Supplier:  Cayman Chemical 146431 G
Encompass_Preferred
A derivative of thalidomide; reduces TNF-α production in isolated human PBMCs and whole blood stimulated by LPS from S. minnesota (IC50s = 13 and 25 nM, respectively); has been used as a building block in the synthesis of PROTACs that induce the degradation of IKZF1 and IKZF3 in MM.1S cells; reduces vascularization and total microvascular length in a rat mesenteric window assay model at 250 mg/kg per day, p.o.
Catalog No. 50-296-9592