Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LENG1 Mouse anti-Human, Clone: OTI1E8, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503113
Description
The function of this protein remains unknown.Specifications
LENG1 | |
Monoclonal | |
0.44 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96BZ8 | |
leukocyte receptor cluster member 1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
79165 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1E8 | |
Unconjugated | |
LENG1 | |
Liquid | |
LENG1 | |
Full length human recombit protein of human LENG1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title