Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LENG1 Mouse anti-Human, Clone: OTI10A7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525595

Catalog No. PIMA525595


Add to cart

Description

Description

The function of this protein remains unknown.
Specifications

Specifications

LENG1
Monoclonal
0.22 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96BZ8
LENG1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79165
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI10A7
Unconjugated
LENG1
Liquid
LENG1
Full length human recombit protein of LENG1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit