Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LENG8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566404
Description
Immunogen sequence: FTVEVYETHAR IALEKGDHEE FNQCQTQLKS LYAENLPGNV GEFTAYRILY YIFTKNSGDI TTELAYLTRE LKADPCVAHA LALRTAWALG NYHRFFRLYC HAPCMSGYLV DKFADRE Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.
LENG8 (Leukocyte Receptor Cluster Member 8) is a Protein Coding gene.
Specifications
LENG8 | |
Polyclonal | |
Unconjugated | |
LENG8 | |
AW049671; KIAA1932; LENG8; leukocyte receptor cluster (LRC) member 8; leukocyte receptor cluster member 8; Leukocyte receptor cluster member 8 homolog; MGC40108; pp13842 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PV6 | |
LENG8 | |
Recombinant Human LENG8. Recombinant protein control fragment (Product #RP-107081). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction