Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin/OB Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody

$321.00 - $493.00

Specifications

Antigen Leptin/OB
Clone 2
Dilution Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020173
View Documents
Novus Biologicals
NBP289345100UL
100 μg
Each for $493.00
Add to cart
 
NB020174
View Documents
Novus Biologicals
NBP28934550UL
50 μg
Each for $321.00
Add to cart
 
Description

Description

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Leptin/OB
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100
Monoclonal
Purified
RUO
Human
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Recombinant Human Leptin/OB Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
2
Flow Cytometry, Immunofluorescence
Unconjugated
Rabbit
Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism
0.2 um filtered solution in PBS
3952
IgG
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit