Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin R Antibody (RM0085-9C29), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121725APC
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Mouse samples. It is validated for Western BlotSpecifications
| Leptin R | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0085-9C29 | |
| Western Blot | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| Mouse recombinant EC-domain of LeptinR | |
| 0.1 mL | |
| Signal Transduction | |
| 3953 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction