Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin R Mouse anti-Human, Alexa Fluor 700, Clone: 52263, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB867N100UG

Catalog No. FAB867N100


Add to cart
This item is not returnable. View return policy

Description

Description

Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Leptin R
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P48357
LEPR
Mouse myeloma cell line NS0-derived recombinant human Leptin R Phe22-Asp839 (predicted) Accession # P48357
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Leptin R in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Leptin R is observed.
Flow Cytometry
52263
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Mouse
IgG2b
RUO
Primary
3953
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)