Learn More
Leptin R Rat anti-Human, Alexa Fluor 350, Clone: 55305, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB497U100UG
Catalog No. FAB497U100U
Add to cart
This item is not returnable. View return policy
Description
Description
Leptin R Monoclonal antibody specifically detects Leptin R in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
Leptin R
55305
Flow Cytometry 0.25-1 ug/106 cells
P48356
B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295
Mouse myeloma cell line NS0-derived recombinant mouse Leptin R, Leu21-Leu840 (predicted), Accession # P48356
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3953
Human
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rat
IgG2a
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Reconstitution default not known