Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin R Rat anti-Mouse, Alexa Fluor 405, Clone: RM0085-9C29, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121725AF405
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.Specifications
Leptin R | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LEPR | |
Mouse recombinant EC-domain of LeptinR | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0085-9C29 | |
Western Blot | |
CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
3953 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title