Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptomycin B, Tocris Bioscience™
Click to view available options
Quantity:
5 μg
Description
Leptomycin B is an antifungal antibiotic that is an inhibitor of the nuclear export of proteins; acts by binding directly to and inhibiting CRM1/exportin-1. Inhibits the nuclear export of the HIV regulatory protein Rev and stabilizes p53. Antitumor in vitro and in vivo.
Specifications
Type | Leptomycin B |
Formula Weight | 540.74 |
Percent Purity | >98% |
Quantity | 5 μg |
Chemical Name or Material | Leptomycin B |
Safety and Handling
Recommended Storage : Desiccate at -20°C