Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lercanidipine hydrochloride, Tocris Bioscience™

Catalog Number 295950
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
132866-11-6
C36H42ClN3O6
648.197
WMFYOYKPJLRMJI-UHFFFAOYSA-N
lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen
157917
5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl
This item is not returnable. View return policy
132866-11-6
C36H42ClN3O6
648.197
WMFYOYKPJLRMJI-UHFFFAOYSA-N
lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen
157917
5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl

CaV1.x blocker

L-type Ca2+ channel blocker that displays higher vascular selectivity than felodipine (Cat. No. 2960). Causes peripheral vasodilation with only weak negative inotropic activity. Antihypertensive. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 2.36 μM).
Quantity 50 mg
Formula Weight 648.19
Percent Purity >99%
Chemical Name or Material Lercanidipine hydrochloride
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit