Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lercanidipine hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
L-type Ca2+ channel blocker that displays higher vascular selectivity than felodipine (Cat. No. 2960). Causes peripheral vasodilation with only weak negative inotropic activity. Antihypertensive. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 2.36 μM).
Specifications
Quantity | 10 mg |
Formula Weight | 648.19 |
Percent Purity | >99% |
Chemical Name or Material | Lercanidipine hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction