Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LETM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552789
Description
Immunogen sequence: STLGLRNCLN VPFGCCTPIH PVYTSSRGDH LGCWALRPEC LRIVSRAPWT STSVGFVAVG PQCLPVRGWH SSRPVRDDSV VEKSLKSL Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 53%.
This gene encodes a protein that is localized to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.
Specifications
LETM1 | |
Polyclonal | |
Unconjugated | |
LETM1 | |
LETM1; LETM1 and EF-hand domain-containing protein 1, mitochondrial; leucine zipper and EF-hand containing transmembrane protein 1; leucine zipper-EF-hand containing transmembrane protein 1; leucine zipper-EF-hand-containing transmembrane protein 1; Mdm38 homolog; Mitochondrial proton/calcium exchanger protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3954 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95202 | |
LETM1 | |
Recombinant protein corresponding to Human LETM1. Recombinant protein control fragment (Product #RP-90426). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction