Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukadherin 1, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
344897-95-6
C22H15NO4S2
421.485
AEZGRQSLKVNPCI-UNOMPAQXSA-N
leukadherin-1,la1,leukadherin 1,rhodanine-furan analogue 14,4-5-5z-3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid,4-5-4-oxo-3-phenylmethyl-2-thioxo-5-thiazolidinylidene methyl-2-furanyl benzoic acid,4-5-z-3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid,4-5-4-oxo-3-phenylmethyl-2-thioxo-5-thiazolidinylidene methyl-2-furanyl-benzoic acid,4-5-z-3-benzyl-4-oxo-2-thioxo-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid
5342077
4-[5-[(Z)-(3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene)methyl]furan-2-yl]benzoic acid
C1=CC=C(C=C1)CN2C(=O)C(=CC3=CC=C(O3)C4=CC=C(C=C4)C(=O)O)SC2=S
This item is not returnable. View return policy
344897-95-6
C22H15NO4S2
421.485
AEZGRQSLKVNPCI-UNOMPAQXSA-N
leukadherin-1,la1,leukadherin 1,rhodanine-furan analogue 14,4-5-5z-3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid,4-5-4-oxo-3-phenylmethyl-2-thioxo-5-thiazolidinylidene methyl-2-furanyl benzoic acid,4-5-z-3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid,4-5-4-oxo-3-phenylmethyl-2-thioxo-5-thiazolidinylidene methyl-2-furanyl-benzoic acid,4-5-z-3-benzyl-4-oxo-2-thioxo-1,3-thiazolidin-5-ylidene methyl furan-2-yl benzoic acid
5342077
4-[5-[(Z)-(3-benzyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene)methyl]furan-2-yl]benzoic acid
C1=CC=C(C=C1)CN2C(=O)C(=CC3=CC=C(O3)C4=CC=C(C=C4)C(=O)O)SC2=S

Allosteric activator of CD11b/CD18

Allosteric activator of CD11b/CD18. Increases CD11b/CD18-dependent cell adhesion to fibrinogen (EC50 = 4μM). Decreases leukocyte motility and transendothelial migration; reduces inflammation. Also activates complement receptor 3 (CR3), and promotes CR3-dependent leukocyte adhesion to fibrinogen-coated surfaces.
Quantity 10mg
Formula Weight 421.49
Percent Purity >98%
Chemical Name or Material Leukadherin 1

Recommended Storage : Store at +4°C