Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukemia Virus Mouse anti-Cat, Clone: 8D11, Abnova™

Mouse monoclonal antibody raised against cat Leukemia Virus.

Manufacturer:  Abnova Corporation MAB12431

Catalog No. 89-934-132


Add to cart

Description

Description

Specifications

Specifications

Leukemia Virus
8D11
Mouse monoclonal antibody raised against cat Leukemia Virus.
In PBS, pH 7.2 (0.09% sodium azide)
Mouse
IgG
1 mg
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Immunoprecipitation, Western Blot
Unconjugated
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Native purified Feline Leukemia Virus.
Antigen Affinity Purification
RUO
Primary
Feline
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit