Learn More
StemExpress Human Peripheral Blood Leukopak, Fresh, Size: 10 billion TNCs
SDP
Supplier:  StemExpress LE010F
Encompass
Leukopaks are collected from healthy IRB consented donors, tested negative for HBV, HCV, and HIV, using the Spectra Optia Apheresis System, where cell-rich mononuclear cells are extracted using a continuous flow system directly into a sterile collection bag containing the anticoagulant ACD-A. For Research Use Only. Not for Diagnostic or Therapeutics use, and are not to be administed to humans.
Catalog No. 50-212-6317
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.