Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene A4 Hydrolase/LTA4H Mouse anti-Human, Rat, Canine, Monkey, Alexa Fluor™ 405, Clone: OTI1E9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271189AF405

Catalog No. N271189A405


Add to cart

Description

Description

Leukotriene A4 Hydrolase/LTA4H Monoclonal antibody specifically detects Leukotriene A4 Hydrolase/LTA4H in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Leukotriene A4 Hydrolase/LTA4H
OTI1E9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
LTA4H
Full-length protein expressed in 293T cell transfected with human Leukotriene A4 Hydrolase/LTA4H expression vector.
Immunogen affinity purified
Lipid and Metabolism
Primary
4048.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit