Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leupeptin, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
5mg
25mg
100mg
Packaging:
Plastic bottle
55123-66-5
C20H38N6O4
426.562
MFCD00037012
GDBQQVLCIARPGH-ULQDDVLXSA-N
leupeptin,ac-leu-leu-arg-h,acetyl-l-leucyl-l-leucylargininal,leupeptin hemisulfate,n-acetyl-l-leucyl-l-leucyl-l-argininal,unii-j97339nr3v,leupeptin ac-ll,acetyl-leu-leu-arg-al,l-leucinamide, n-acetyl-l-leucyl-n-4-aminoiminomethyl amino-1-formylbutyl,s-2-2-acetamido-4-methylvaleramido-n-1-formyl-4-guanidinobutyl-4-methyl-valeramide
72429
(2S)-2-acetamido-N-[(2S)-1-[[(2S)-5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]-4-methylpentanamide
CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O)NC(=O)C
55123-66-5
C20H38N6O4
426.562
MFCD00037012
GDBQQVLCIARPGH-ULQDDVLXSA-N
leupeptin,ac-leu-leu-arg-h,acetyl-l-leucyl-l-leucylargininal,leupeptin hemisulfate,n-acetyl-l-leucyl-l-leucyl-l-argininal,unii-j97339nr3v,leupeptin ac-ll,acetyl-leu-leu-arg-al,l-leucinamide, n-acetyl-l-leucyl-n-4-aminoiminomethyl amino-1-formylbutyl,s-2-2-acetamido-4-methylvaleramido-n-1-formyl-4-guanidinobutyl-4-methyl-valeramide
72429
(2S)-2-acetamido-N-[(2S)-1-[[(2S)-5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]-4-methylpentanamide
CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCCN=C(N)N)C=O)NC(=O)C
  • Protease inhibitor, will strongly inhibit Trypsin, Papain, Plasmin, Thrombokinase, Kallikrein and Cathepsin B
  • Does not inhibit Chymotrypsin, Elastase, Renin, or Pepsin
Packaging Plastic bottle
Quantity 5mg
Formula Weight 475.59
Physical Form Powder
Grade Ultrapure
Chemical Name or Material Leupeptin