Learn More
Medchemexpress LLC Levacetylleucine | 1188-21-2 | ≥98.0% | C8H15NO3 | 100 G
Supplier: Medchemexpress LLC HY59291100G
Levacetylleucine is an orally bioavailable and brain-penetrant compound, and an acetylated derivative of the amino acid Leucine. It represents the active form of N-acetyl-leucine (NAL). This compound has been shown to attenuate neuronal death and neuroinflammation and may improve lysosomal and metabolic dysfunction.
- Orally bioavailable and brain-penetrant
- Acetylated derivative of amino acid leucine
- Attenuates neuronal death and neuroinflammation
- Potentially improves lysosomal and metabolic dysfunction
- Improves compensation of postural symptoms
- Promising for research of Niemann-Pick disease type C
- Promising for research of traumatic brain injury
- Promising for research of neurodegeneration prevention