Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levamisole Hydrochloride, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
16595-80-5
C11H13ClN2S
240.75
LAZPBGZRMVRFKY-HNCPQSOCSA-N
hydrogen (6S)-6-phenyl-2H,3H,5H,6H-imidazo[2,1-b][1,3]thiazole chloride
[H+].[Cl-].C1CN2C[C@@H](N=C2S1)C1=CC=CC=C1
16595-80-5
C11H13ClN2S
240.75
LAZPBGZRMVRFKY-HNCPQSOCSA-N
hydrogen (6S)-6-phenyl-2H,3H,5H,6H-imidazo[2,1-b][1,3]thiazole chloride
[H+].[Cl-].C1CN2C[C@@H](N=C2S1)C1=CC=CC=C1

L1054, 16595-80-5, C11H12N2S•HCl

Spectrum™ Levamisole Hydrochloride, USP is used as an immunomodulator and anthelmintic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 25 kg
Specific Rotation -1°21.5to-1°28.0°
Melting Range 226°-231°C
CAS Max % 1
UN Number UN2811
Percent Purity 98.5 to 101%