Learn More
Level 2 SMS Lab Coat
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- SMS Non-woven material
- Knitting and elastic cuff for more protection
- Fluid resistant
- Color: white
- AAMI Level 2 standard
Specifications
Type | Single-use Lab Coat |
Size | Large |
Material | SMS Fabric, SMS |
Color | White |
Cuff Style | Knit |
Sterility | Non-sterile |
Weight (Metric) | 37 gsm |