Learn More
LevGo smartSpatula™ Disposable Polypropylene Spatula, Green Products for Science Education
smartSpatulas offer an economical, disposable tool for transferring, collecting, and handling chemical and biological materials
Manufacturer:  Levgo Inc 17241/EMD
Catalog No. S28422
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Economical, Polypropylene, Single-Use Laboratory Spatulas
  • Ideal for Transferring, Manipulating, and Mixing Materials
  • Versatile Double-Ended Design
  • Help Minimize Cross Contamination
  • Resistant to acids and bases
  • Autoclavable
Specifications
Specifications
Yes
Spatula, Macro
14mm
Green
Use as a spatula, spoon, scoop, scraper, stirring rod, or policeman
310mm