Alfa Aesar™ Levulinic acid, 98%

Manufacturer: Alfa Aesar™ A108130I

Quantity 5000g

 View more versions of this product

Catalog No. AAA108130I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123-76-2
    Molecular Formula C5H8O3
    Formula Weight 116.12
    MDL Number MFCD00002796
    UN Number UN3261
    Beilstein 506796
    Merck Index 14,5472
    Synonym 4-Ketopentanoic acid; 4-Oxopentanoic acid
    Chemical Name or Material Levulinic acid

    Specifications

    Formula Weight 116.12
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Odor Characteristic
    Beilstein 506796
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Quantity 5000g
    Molecular Formula C5H8O3
    Merck Index 14,5472
    Health Hazard 1 Danger
    Melting Point 26° to 31°C
    Boiling Point 245° to 246°C
    Flash Point 137°C (278°F)
    TSCA TSCA
    MDL Number MFCD00002796
    CAS 123-76-2
    UN Number UN3261
    Synonym 4-Ketopentanoic acid; 4-Oxopentanoic acid
    pH 2.5 to 3.0
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number OI1575000
    Chemical Name or Material Levulinic acid
    Assay 98%

    Description

    Applications
    Mainly used as an intermediate for medicines, agricultural chemical and organic synthesis. Used as raw material of perfume, modifier and solvent of plastics. It is also employed as an additive of polymer, paints and lubricating oil. Also used as a surfactant and auxiliary for making printing ink, rubber flux and cosmetics(including shampoo).

    Solubility
    Soluble in water ( 675g/L at 20°C).

    Notes
    Light Sensitive. Store in dark. Store away from oxidizing agents and bases.

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit