Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levulinic Acid, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
123-76-2
C5H8O3
116.12
MFCD00002796
JOOXCMJARBKPKM-UHFFFAOYSA-N
levulinic acid,laevulinic acid,pentanoic acid, 4-oxo,levulic acid,4-oxovaleric acid,3-acetylpropionic acid,4-ketovaleric acid,leva,gamma-ketovaleric acid,acetopropionic acid
11579
CHEBI:45630
4-oxopentanoic acid
CC(=O)CCC(=O)O
123-76-2
C5H8O3
116.12
MFCD00002796
JOOXCMJARBKPKM-UHFFFAOYSA-N
levulinic acid,laevulinic acid,pentanoic acid, 4-oxo,levulic acid,4-oxovaleric acid,3-acetylpropionic acid,4-ketovaleric acid,leva,gamma-ketovaleric acid,acetopropionic acid
11579
CHEBI:45630
4-oxopentanoic acid
CC(=O)CCC(=O)O
Infrared Spectrum Authentic
Density 1.1300g/mL
Flash Point 98°C
Quantity 250g
Refractive Index 1.4396 to 1.4426
Packaging Glass bottle
Boiling Point 245.0°C to 246.0°C
Specific Gravity 1.13
Melting Point 33.0°C to 35.0°C
Water 1% max. (K.F.)
Color Yellow
Assay Percent Range 98+%
Linear Formula CH3COCH2CH2CO2H
Beilstein 03, 671
Fieser 01,564; 02,239; 10,230
Merck Index 15, 5526
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and ether,insoluble in aliphatic hydrocarbons
Formula Weight 116.12
Physical Form Liquid After Melting
Percent Purity ≥98%
Chemical Name or Material Levulinic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger