Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Lewatit™ MonoPlus M 500, Chloride Form, MilliporeSigma™ Supelco™
Specifications
Product Line Lewatit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-5056
View Documents
MilliporeSigma Supelco
62096250GF
250 g
Each for $48.45
N/A
11-100-4409
View Documents
MilliporeSigma Supelco
620961KGF
1 kg
Each for $156.90
N/A
Description
Description
Provides the filter bed with special properties: high exchange rate, very high utilization of the total capacity, low amount of washing water needed, almost linear skin-friction gradient for the entire height of bed, very good separation of the components in the mixed-bed filter.; anion exchange resin of gel type; spherical particles of the same diameter (monodispers)
Specifications
Specifications
Lewatit
SDS
Product Certifications
Lewatit is a registered trademark of Lanxess Deutschland GmbH