Learn More
Lewatit™ MP-62, Free Base, MilliporeSigma™ Supelco™
Lewatit™ MP-62 is a weakly basic anionic macroporous resin which has tertiary amine as functional group. It is applicable in noble metal recovery like platinum (VI) sorption, palladium(II) removal.
$33.00 - $91.80
Specifications
Product Line Lewatit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-5152
View Documents
MilliporeSigma Supelco
62088100G
100 g
Each for $33.00
Add to cart
11-100-4662
View Documents
MilliporeSigma Supelco
62088500G
500 g
Each for $91.80
Add to cart
Description
Description
Specifications
Specifications
Lewatit
SDS
Product Certifications
Lewatit is a registered trademark of Lanxess Deutschland GmbH