Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewatit™ TP 207, Sodium Form, MilliporeSigma™ Supelco™

$37.80 - $143.80

Specifications

CAS 57285-14-0
Product Line Lewatit
Physical Form Particles (spherical)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4698
View Documents
MilliporeSigma Supelco
62103100G
100 g
Each for $37.80
Add to cart
 
11-101-4697
View Documents
MilliporeSigma Supelco
62103500G
500 g
Each for $143.80
Add to cart
 
Description

Description

Specifications

Specifications

57285-14-0
Lewatit
Particles (spherical)
SDS
Product Certifications

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit