Learn More
Lewis Y (Blood Group Antigen) Mouse anti-Human, Clone: A70-C/C8, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB15084
Catalog No. 89-035-104
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse monoclonal antibody raised against native human Lewis Y (Blood Group Antigen).
  • High specificity
  • High reactivity
Specifications
Specifications
Lewis Y (Blood Group Antigen)
A70-C/C8
Mouse monoclonal antibody raised against native human Lewis Y (Blood Group Antigen).
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Mouse
IgM
100 ug
Store at 4°C.
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Live Ls174T cells (human colon carcinoma cell line).
PEG precipitation
RUO
Primary
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)