Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewis Y Mouse anti-Human, PE/Cy5.5, Clone: LWY/1463, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254560PECY55

Catalog No. N254560PEC5


Add to cart

Description

Description

Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Lewis Y
LWY/1463
Flow Cytometry
Conjugated, Purified
Human colon carcinoma cells
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Mouse
IgG1 Kappa
0.1 mL
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit