Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lewis Y, Rabbit anti-Human, Clone: BR96, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810770.2MG
Description
Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Lewis Y | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry | |
Rabbit | |
Protein A purified | |
RUO | |
The antibody binds Lewis Y with a Ka of 0.1 uM. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
ELISA, Immunohistochemistry | |
BR96 | |
Unconjugated | |
PBS | |
H3396 cultured line established from a metastasis of a human breast adenocarcinoma. | |
0.2 mg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction