Learn More
3M™ Lexa™ GoggleGear™
Wider view than traditional impact goggles
Supplier:  3M™ 7100075727
 View more versions of this product
Catalog No. 19-145-423
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
This item is not returnable. View return policy
Description
Description
  • Clear lens absorbs 99.9% UV
  • Top and bottom vent channels increase airflow, minimize fogging
  • A soft foam flange seals the entire lens perimeter to help keep out particulates
  • Comfortable enough to wear all day
  • 3M Catalog# 16617-00000-10 , 3M Catalog# 16618-00000-10
Order Info
Available with adjustable temple for individualized fit or headband for secure fit in active applications.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
Specifications
Specifications
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
Clear
Black
16617-00000-10
Anti-fog hardcoat
Anti-fog
Safety goggles
Foam backing seals, Top and bottom vent channels, headband frame
Medium