Learn More
3M™ Lexa™ GoggleGear™
Wider view than traditional impact goggles
Supplier: 3M™ 7010045732
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Description
- Clear lens absorbs 99.9% UV
- Top and bottom vent channels increase airflow, minimize fogging
- A soft foam flange seals the entire lens perimeter to help keep out particulates
- Comfortable enough to wear all day
- 3M Catalog# 16617-00000-10 , 3M Catalog# 16618-00000-10
Order Info
Available with adjustable temple for individualized fit or headband for secure fit in active applications.
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
Specifications
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards | |
Clear | |
Black | |
16618-00000-10 | |
Anti-fog hardcoat |
Anti-fog | |
Safety goggles | |
Foam backing seals, Top and bottom vent channels | |
Large |