LG 100754, Tocris™

RXR:PPAR agonist

Manufacturer: R&D Systems 3831/10

Quantity 10mg

 View more versions of this product

Catalog No. 38-311-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C26H36O3
    Formula Weight 396.56
    Synonym (2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
    Chemical Name or Material LG 100754

    Specifications

    Formula Weight 396.56
    Quantity 10mg
    Synonym (2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
    Molecular Formula C26H36O3
    CAS 180713-37-5
    Assay Percent Range >99%
    Recommended Storage Store at -20°C
    Chemical Name or Material LG 100754

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit