Learn More
Electron Microscopy SciencesSupplier Diversity Partner Large Paraffin Dispenser, 50/60Hz, 2.25 Amp, 220/240V DFS Item
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 6284020
Holds 6.25 gallons of paraffin wax with clear lexan lid permits easy viewing of paraffin without contamination. High spigot accommodates large transfer vessels. The unit comes with microprocessor temperature regulation, built-in clock. Ambient to 65 degree C.The Paraffin Dispenser have been tested by a registered independent body to rigorous international quality and safety standards, and, have been found to be in compliance with UL, CSA and CE.
Catalog No. 50-190-0628